BAY 11-7085 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (E)-3-(4-tert-Butylphenylsulfonyl)acrylonitrile(E)-3-((4-(tert-Butyl)phenyl)sulfonyl)acrylonitrile(E)-3-(4-tert-Butylphenylsulphonyl)acrylonitrile(E)-3-((4-(tert-Butyl)phenyl)sulphonyl)acrylonitrile

J67222-MA
AAJ67222-MAEA 169 USD
AAJ67222-MA
BAY 11-7085 99%
BAY 11-7085

Solid

Formula: C₁₃H₁₅NO₂S
MW: 249.328 g/mol
Melting Pt: 80…82 °C
Storage Temperature: Ambient
MDL Number: MFCD01862602
CAS Number: 196309-76-9

Order Now

Specification Test Results

Assay Minimum 99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR