Gold carboxy-functionalized, 3kDa PEGylated, nanoparticles 15 nm OD50 (Abs. 520 nm)

J67217-XF J67217-AMI
AAJ67217-AMIEA 253.98 USD
AAJ67217-AMI AAJ67217-XF
Gold carboxy-functionalized, 3kDa PEGylated, nanoparticles 15 nm OD50 (Abs. 520 nm)
Gold
Formula: Au
MW: 196.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR