Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilized, nanoparticles 400 nm

J67213-AC J67213-AE
AAJ67213-ACEA 193.4 USD
AAJ67213-AC AAJ67213-AE
Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilized, nanoparticles 400 nm
Gold

Liquid

Formula: Au
MW: 196.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR