Gold supplied in purified water biotin-functionalized, PEGylated, nanoparticles 60 nm OD50 (Abs. 540 nm)

J67210-AMI J67210-XF
AAJ67210-AMIEA 253.98 USD
AAJ67210-AMI AAJ67210-XF
Gold supplied in purified water biotin-functionalized, PEGylated, nanoparticles 60 nm OD50 (Abs. 540 nm)
Gold
Formula: Au
MW: 196.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR