Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formamide ≥99.5%, ultrapure

Supplier: Alfa Aesar
Danger
Synonyms: Carbamaldehyde

J67206-AE J67206-AK
AAJ67206-AEEA 45.97 USD InStock
AAJ67206-AE AAJ67206-AK
Formamide ≥99.5%, ultrapure
Formamide
Liquid


Formula: HCONH₂
MW: 45.04 g/mol
Boiling Pt: 210 °C
Melting Pt: 2 °C
Density: 1,134
Flash Pt: 150 °C (302 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007941
CAS Number: 75-12-7
EINECS: 200-842-0
Merck Index: 13,04261
https://us.vwr.com/stibo/web/std.lang.all/25/02/2302502.jpg
Order Now
Specification Test Results
Assay Minimum 99.5

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty