Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tyrphostin RG 14620 ≥98%

Supplier: Alfa Aesar

J67185-MA J67185-LB0
AAJ67185-MAEA 146.18 USD InStock
AAJ67185-MA AAJ67185-LB0
Tyrphostin RG 14620 ≥98%
Tyrphostin RG 14620
Solid


Formula: C₁₄H₈Cl₂N₂
MW: 275.132 g/mol
Melting Pt: 150-151°C
Storage Temperature: Ambient
MDL Number: MFCD00236445
CAS Number: 136831-49-7
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty