Citrate Buffer, 0.2M Buffer Soln., pH 5.0, Low Endotoxin, Thermo Scientific

Supplier: Thermo Scientific
J67180-AE J67180-AK
AAJ67180-AKEA 103.73 USD
AAJ67180-AK AAJ67180-AE
Citrate Buffer, 0.2M Buffer Soln., pH 5.0, Low Endotoxin, Thermo Scientific
Buffers Cell Culture Buffers
Liquid

CAS No.: 6132-04-3
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00150031
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR