Silver nanoparticles supplied in 2 mM sodium citrate 10 nm, conjugated to Protein A

J67161-XF
AAJ67161-XFEA 253.98 USD
AAJ67161-XF
Silver nanoparticles supplied in 2 mM sodium citrate 10 nm, conjugated to Protein A
Silver
Formula: Ag
MW: 107.87 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR