N,N,2-Trimethyl-5-nitrobenzenesulfonamide 98%

Synonyms: BRL 50481

J67146-MA
AAJ67146-MAEA 157.44 USD
AAJ67146-MA
N,N,2-Trimethyl-5-nitrobenzenesulfonamide 98%
N,N,2-Trimethyl-5-nitrobenzenesulfonamide

Solid

Formula: C₉H₁₂N₂O₄S
MW: 244.27 g/mol
Melting Pt: 65…67 °C
Storage Temperature: Ambient
MDL Number: MFCD03039912
CAS Number: 433695-36-4

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR