Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gold supplied in 0.1 mM PBS ≥99%, nanoparticles 300 nm reactant-free

Supplier: Alfa Aesar

J67137-AE J67137-AC
AAJ67137-AEEA 640.84 USD InStock
AAJ67137-AE AAJ67137-AC
Gold supplied in 0.1 mM PBS ≥99%, nanoparticles 300 nm reactant-free
Gold
Liquid


Formula: Au
MW: 196,97 g/mol
Boiling Pt: 2800 °C (1013 hPa)
Melting Pt: 1063 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526
Order Now
Specification Test Results
Assay Minimum 99

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty