N-(4-Phenylsulfonylphenyl)-3,3,3-trifluoro-2-hydroxy-2-methylpropanamide 98%

Warning

Synonyms: ZM-226600

J67133-MA
AAJ67133-MAEA 186.47 USD
AAJ67133-MA
N-(4-Phenylsulfonylphenyl)-3,3,3-trifluoro-2-hydroxy-2-methylpropanamide 98%
N-(4-Phenylsulfonylphenyl)-3,3,3-trifluoro-2-hydroxy-2-methylpropanamide

Solid

Formula: C₁₆H₁₄F₃NO₄S
MW: 373.346 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09266222
CAS Number: 147695-92-9

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR