Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Imidazole 99+% Molecular biology grade

Supplier: Alfa Aesar
Danger
Synonyms: Glyoxaline 1H-Imidazole Glyoxalamine

J67132-49 J67132-22 J67132-6Y J67132-14 J67132-18
AAJ67132-49PK 382.44 USD InStock
AAJ67132-49 AAJ67132-22 AAJ67132-6Y AAJ67132-14 AAJ67132-18
Imidazole 99+% Molecular biology grade
Imidazole
Powder


Formula: C₃H₄N₂
MW: 68.08 g/mol
Boiling Pt: 256°C
Melting Pt: 90°C
Density: 1.03
Flash Pt: 145°C (293°F)
Storage Temperature: Ambient
MDL Number: MFCD00005183
CAS Number: 288-32-4
EINECS: 206-019-2
UN: 3263
ADR: 8,III
Merck Index: 13,04935
https://us.vwr.com/stibo/web/std.lang.all/84/13/2298413.jpg
Order Now
Specification Test Results
Assay Minimum 99

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty