17-Phenyltrinorprostaglandin E₂ ≥98%

Danger

J67126-MCR
AAJ67126-MCREA 171.63 USD
AAJ67126-MCR
17-Phenyltrinorprostaglandin E₂ ≥98%
17-Phenyltrinorprostaglandin E2

Oil

Formula: C₂₃H₃₀O₅
MW: 386.488 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00135219
CAS Number: 38315-43-4

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR