Bopindolol malonate ≥97%
Supplier: Thermo Scientific Chemicals
Synonyms:
HE393689
, AC1LCS45
, CTK8F831
, SCHEMBL348872
, D01794
, Wandonorm
, LS-120081
J67119-MC
AAJ67119-MCEA
142.02
USD
AAJ67119-MC
Bopindolol malonate ≥97%
Bopindolol malonate
A slowly dissociating _x001A_1-adrenergic receptor antagonist that has antioxidant and free radical scavenging activity.
Solid
Formula:
C₂₇H₃₂N₂O₇ MW: 484.54 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00798228 CAS Number: 62658-64-4 |
Specification Test Results
Assay Minimum | 99 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...