Gold 99% supplied in 0.1 mM PBS, nanoparticles 30 nm reactant-free OD1 (Abs. 526 nm)
Supplier: Thermo Scientific Chemicals
J67118-AE
J67118-AC
AAJ67118-AEEA
362.05
USD
AAJ67118-AE
AAJ67118-AC
Gold 99% supplied in 0.1 mM PBS, nanoparticles 30 nm reactant-free OD1 (Abs. 526 nm)
Gold
Ideal for sensitive applications requiring a minimum amount of contaminating reactants. Well suited for protein conjugate development by passive adsorption to the gold nanoparticle surface. Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications
Liquid
Formula:
Au MW: 196.97 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 |
Specification Test Results
Assay Minimum | 99 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...