Methyl-2-(4-fluorobenzamido)benzoate 98%

Warning

J67103-MA
AAJ67103-MAEA 141.43 USD
AAJ67103-MA
Methyl-2-(4-fluorobenzamido)benzoate 98%
Methyl-2-(4-fluorobenzamido)benzoate

Solid

Formula: C₁₅H₁₂FNO₃
MW: 273.263 g/mol
Melting Pt: 132…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00028104
CAS Number: 461681-88-9

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR