1-O-Palmitoyl-2-O-acetyl-sn-glycero-3-phosphocholine ≥99%

J67083-MCR J67083-LB0
AAJ67083-LB0EA 429.91 USD
AAJ67083-LB0 AAJ67083-MCR
1-O-Palmitoyl-2-O-acetyl-sn-glycero-3-phosphocholine ≥99%
1-O-Palmitoyl-2-O-acetyl-sn-glycero-3-phosphocholine

Viscous oil

Formula: C₂₆H₅₂NO₈P
MW: 537.675 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00798242
CAS Number: 84062-61-3

Order Now

Specification Test Results

Assay Minimum 99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR