Isotetrandrine ≥98%
Supplier: Thermo Scientific Chemicals
J67081-MCR
AAJ67081-MCREA
123.8
USD
AAJ67081-MCR
Isotetrandrine ≥98%
Isotetrandrine
Solid
Formula:
C₃₈H₄₂N₂O₆ MW: 622.762 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00236404 CAS Number: 477-57-6 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...