Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-myo-Inositol-1,5,6-trisphosphate sodium salt

Supplier: Alfa Aesar
Synonyms: Ins(1,5,6)-P3 sodium salt1,5,6-IP3 sodium salt

J67076-EXD J67076-EXZ
AAJ67076-EXDEA 418.25 USD InStock
AAJ67076-EXD AAJ67076-EXZ
D-myo-Inositol-1,5,6-trisphosphate sodium salt
D-myo-Inositol-1,5,6-trisphosphate sodium salt
Lyophilized powder


Formula: C₆H₁₂Na₃O₁₅P₃
MW: 486.04 g/mol
Storage Temperature: Freezer
MDL Number: MFCD12912676
CAS Number: 120965-76-6
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty