Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilized, nanoparticles 300 nm

Supplier: Thermo Scientific Chemicals

J67066-AE J67066-AC
AAJ67066-ACEA 185.54 USD
AAJ67066-AC AAJ67066-AE
Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilized, nanoparticles 300 nm
Gold

For adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.


Liquid

Formula: Au
MW: 196.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR