Fumagillin ≥95%

Supplier: Thermo Scientific Chemicals
Warning

J67045.MCR
AAJ67045-MCREA 219.1 USD
AAJ67045-MCR
Fumagillin ≥95%
Fumagillin

Solid

Formula: C₂₆H₃₄O₇
MW: 458.551 g/mol
Melting Pt: 194…195 °C
Storage Temperature: Freezer
MDL Number: MFCD03990453
CAS Number: 23110-15-8
EINECS: 245-433-8

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR