Tyrphostin AG-1296 ≥98%

Synonyms: 6,7-Dimethoxy-2-phenylquinoxaline

J67039-LB0
AAJ67039-LB0EA 237.02 USD
AAJ67039-LB0
Tyrphostin AG-1296 ≥98%
Tyrphostin AG-1296

Solid

Formula: C₁₆H₁₄N₂O₂
MW: 266.3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00270913
CAS Number: 146535-11-7

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR