LY 171883 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tomelukast

J67028-MA
AAJ67028-MAEA 172.47 USD
AAJ67028-MA
LY 171883 98%
LY 171883

Solid

Formula: C₁₆H₂₂N₄O₃
Melting Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00211057
CAS Number: 88107-10-2

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR