Gö 6983 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-[1-[3-(dimethylamino)propyl]-5-methoxy-1H-indol-3-yl]-4-(1H-indol-3-yl)-1H-pyrrole-2,5-dione

J67008-MA J67008-MCR J67008-LB0
AAJ67008-MAEA 575.35 USD
AAJ67008-MA AAJ67008-MCR AAJ67008-LB0
Gö 6983 98%
Gö 6983

Solid

Formula: C₂₆H₂₆N₄O₃
MW: 442.519 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04040031
CAS Number: 133053-19-7

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR