CAPS, 0.2M Buffer Soln., pH 9.0, Low Endotoxin, Thermo Scientific

Supplier: Thermo Scientific
J67007-AE J67007-AK
AAJ67007-AKEA 85.73 USD
AAJ67007-AK AAJ67007-AE
CAPS, 0.2M Buffer Soln., pH 9.0, Low Endotoxin, Thermo Scientific
Buffers Cell Culture Buffers
Liquid

CAS No.: 1135-40-6
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00003837
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR