Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Albumin, human, 96%

Supplier: Alfa Aesar
J66780-06 J66780-03 J66780-ME
AAJ66780-06EA 184.9 USD InStock
AAJ66780-06 AAJ66780-03 AAJ66780-ME
Albumin, human, 96%
Proteins and Peptides
Lyophilized powder

Purity: 96%
CAS No.: 70024-90-7
Storage Temperature: 2°C to 8°C
Physical Form: Lyophilized powder
Appearance: White to pale brown
MDL No.: MFCD00081418

Applications: Produced in the liver, albumin is the most abundant protein in blood plasma. A carrier protein for steroids, fatty acids, and thyroid hormones, and is vital in regulating the colloidal osmotic pressures of blood. Binds exogenous substances.

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty