Alpha-Conotoxin MI

Supplier: Thermo Scientific
J66753-MCR J66753-EXD
AAJ66753-MCREA 169.51 USD
AAJ66753-MCR AAJ66753-EXD
Alpha-Conotoxin MI
Proteins and Peptides
Lyophilized powder

Molecular Formula: C58H88N22O17S4
Formula Weight: 1493.71
Storage Temperature: 2°C to 8°C
Physical Form: Lyophilized powder
Appearance: White to off-white
MDL No.: MFCD00151773
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR