Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

alpha-Conotoxin SIA

Supplier: Alfa Aesar
J66745-MCR J66745-EXD
AAJ66745-MCREA 250.62 USD InStock
AAJ66745-MCR AAJ66745-EXD
alpha-Conotoxin SIA
Proteins and Peptides
Lyophilized powder

Molecular Formula: C60H82N18O17S4
Formula Weight: 1455.66
Storage Temperature: 2°C to 8°C
Physical Form: Lyophilized powder
Appearance: White to off-white
MDL No.: MFCD00236931

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty