Sulfamerazine sodium salt

Supplier: Thermo Scientific Chemicals
Warning

J66717-30
AAJ66717-30EA 152.16 USD
AAJ66717-30
Sulfamerazine sodium salt
Sulfamerazine sodium salt

Powder

Formula: C₁₁H₁₁N₄NaO₂S
MW: 286.285 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00068332
CAS Number: 127-58-2
EINECS: 204-851-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR