Potassium peroxodisulfate 99.0% for electrophoresis
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dipotassium peroxodisulphate
J66672-22
J66672-14
AAJ66672-22EA
43.64
USD
AAJ66672-22
AAJ66672-14
Potassium peroxodisulfate 99.0% for electrophoresis
Potassium peroxodisulphate
Crystalline
Formula:
K₂S₂O₈ MW: 270.32 g/mol Melting Pt: 100 °C (dec.) Density: 2.477 Storage Temperature: Ambient |
MDL Number:
MFCD00011386 CAS Number: 7727-21-1 EINECS: 231-781-8 UN: 1492 ADR: "5.1",III |
Specification Test Results
Assay Minimum | 99 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...