Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl sulfoxide, Bioreagent

Supplier: Alfa Aesar
Warning
Synonyms: DMSO

J66650-AE J66650-AD J66650-AP J66650-AK J66650-K2
AAJ66650-AEEA 68.44 USD InStock
AAJ66650-AE AAJ66650-AD AAJ66650-AP AAJ66650-AK AAJ66650-K2
Dimethyl sulfoxide, Bioreagent
Dimethyl sulphoxide
Liquid

Applications: Suitable for biotechnology applications such as peptide and oligonucleotide synthesis. This product is not sterile.


Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189 °C
Melting Pt: 18,4 °C
Density: 1,101
Flash Pt: 87 °C (189 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
UN: 0000
Merck Index: 13,03285
https://us.vwr.com/stibo/web/std.lang.all/77/31/2297731.jpg
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty