Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

alpha-Conotoxin IMI

Supplier: Alfa Aesar
J66642-MCR J66642-EXD
AAJ66642-MCREA 150.6 USD InStock
AAJ66642-MCR AAJ66642-EXD
alpha-Conotoxin IMI
Proteins and Peptides
Lyophilized powder

CAS No.: 156467-85-5
Molecular Formula: C52H78N20O15S4
Formula Weight: 1351.56
Storage Temperature: 2°C to 8°C
Physical Form: Lyophilized powder
Appearance: White to off-white
MDL No.: MFCD00283465

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty