Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-myo-Inositol-1,2-bisphosphate sodium salt

Supplier: Alfa Aesar

J66420-EXZ J66420-EXD J66420-MCR
AAJ66420-EXZEA 129.71 USD InStock
AAJ66420-EXZ AAJ66420-EXD AAJ66420-MCR
D-myo-Inositol-1,2-bisphosphate sodium salt
D-myo-Inositol-1,2-bisphosphate sodium salt
Lyophilized powder


Formula: C₆H₁₂Na₂O₁₂P₂
MW: 384.077 g/mol
Storage Temperature: Freezer
MDL Number: MFCD10567083
CAS Number: 208584-51-4
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty