Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium methotrexate ≥98.5%

Supplier: Alfa Aesar
Danger
Synonyms: Methotrexate disodium salt

J66364-MD
AAJ66364-MDEA 190.42 USD InStock
AAJ66364-MD
Sodium methotrexate ≥98.5%
Sodium methotrexate
Powder

Applications: Inhibits the metabolism of folic acid


Formula: C₂₀H₂₀N₈Na₂O₅
MW: 498.411 g/mol
Boiling Pt: 823 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00274560
CAS Number: 7413-34-5
EINECS: 231-022-0
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Assay Minimum 98.5

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty