Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

alpha-Conotoxin EI, Alfa Aesar

Supplier: Alfa Aesar
J66166-EXG
AAJ66166-EXGEA 154.98 USD InStock
AAJ66166-EXG
alpha-Conotoxin EI, Alfa Aesar
Proteins and Peptides
Lyophilized powder

Molecular Formula: C83H123N26O28S5
Formula Weight: 2093.34
Storage Temperature: 2°C to 8°C
Physical Form: Lyophilized powder
Appearance: White to off-white
MDL No.: MFCD00798708

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty