Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ascorbate oxidase, from Cucurbita sp.

Supplier: Alfa Aesar
J65782-4I J65782-4I
AAJ65782-4IEA 58.99 USD InStock
AAJ65782-4I CAAAJ65782-4I
Ascorbate oxidase, from Cucurbita sp.
Enzymes
Lyophilized powder

CAS No.: 9029-44-1
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: Pale blue
MDL No.: MFCD00130560

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty