Human Native DiI-Lipoprotein (from Plasma)

Supplier: Thermo Scientific
J65568-MCR
AAJ65568-MCREA 978.26 USD
AAJ65568-MCR
Human Native DiI-Lipoprotein (from Plasma)
Liquid

Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR