Lipoprotein Deficient Serum, Human, BT-931, Alfa Aesar
J65516-AC
AAJ65516-ACEA 333.52 USD
AAJ65516-AC
Lipoprotein Deficient Serum, Human, BT-931, Alfa Aesar
Lipoprotein Deficient Serum human is used in cell culture medium
  • Stable for 6 to 12 months if frozen
  • Incompatible with strong oxidizing agents
Each lot is evaluated in culture with our LDLs to determine the degree of up regulation of LDL receptors.
It inhibits the transfer of unesterified cholesterol from low density lipoprotein to high density lipoprotein. Further, it is useful as media supplement for lipoprotein uptake and metabolism studies.