Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DiI-Lipoprotein, high density, human plasma, BT-915

Supplier: Alfa Aesar
J65503-MCR J65503-MCR J65503-AMH
AAJ65503-MCREA 840.96 USD InStock
AAJ65503-MCR CAAAJ65503-MCR AAJ65503-AMH
DiI-Lipoprotein, high density, human plasma, BT-915
Proteins and Peptides
Liquid

Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Sensitivity: Light Sensitive

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty