Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Coenzyme A trilithium salt ≥95%

Supplier: Alfa Aesar

J65434-MC J65434-MC
AAJ65434-MCEA 224.19 USD InStock
CAAAJ65434-MC AAJ65434-MC
Coenzyme A trilithium salt ≥95%
Coenzyme A trilithium salt
Powder

Applications: Useful for fatty acid synthesis and oxidation


Formula: C₂₁H₃₃N₇O₁₆P₃SLi₃
MW: 785.33 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00075848
CAS Number: 18439-24-2
EINECS: 242-317-9
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty