Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphatase Inhibitor Cocktail A

Supplier: Alfa Aesar
J65354-LQ J65354-LQ
AAJ65354-LQEA 137.13 USD InStock
CAAAJ65354-LQ AAJ65354-LQ
Phosphatase Inhibitor Cocktail A
Cell Culture Supplements and Additives
Liquid

Storage Temperature: -30°C to -10°C
Physical Form: Liquid
Appearance: Clear

Applications: For use with tissue and cell extracts, including extracts containing detergents

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty