Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Biliverdin dihydrochloride

Supplier: Alfa Aesar

J65226-MC J65226-MD J65226-MD J65226-MC
AAJ65226-MDEA 497.1 USD InStock
CAAAJ65226-MC AAJ65226-MD CAAAJ65226-MD AAJ65226-MC
Biliverdin dihydrochloride
Biliverdin dihydrochloride
Biliverdin dihydrochloride was found responsible for cytoprotection and induce tolerance to cardiac allografts.

It may regulate the cellular heme degradation process by occupying the heme binding site on heme oxygenase which would prevent access of the substrate to the catalytic site of the enzyme.

Caution: Light sensitive.


Formula: C₃₃H₃₆Cl₂N₄O₆
MW: 655.573 g/mol
Storage Temperature: Freezer
MDL Number: MFCD08669554
CAS Number: 55482-27-4
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty