Would you like to log in again?
Lipoprotein Deficient Serum, Bovine, BT-907, Alfa Aesar
Supplier: Alfa Aesar
J65182-AMG
AAJ65182-AMGEA 397.45 USD InStock
AAJ65182-AMG
Lipoprotein Deficient Serum, Bovine, BT-907, Alfa Aesar
Sera (Serum)
Lipoprotein Deficient Serum, bovine is used for blocking of interfering antibodies
  • Prepared from calf serum
  • Stable for 6 to 12 months if frozen
It is evaluated in conjunction with low-density lipoprotein (LDL) to determine the degree of up regulation of low-density lipoprotein receptors.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty