Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Azadibenzocyclooctyne-PEG, M.W. 5 kDa

Supplier: Alfa Aesar
Warning
Synonyms: ADIBO-PEG

J64977-MC J64977-MC
AAJ64977-MCEA 463.12 USD InStock
CAAAJ64977-MC AAJ64977-MC
Azadibenzocyclooctyne-PEG, M.W. 5 kDa
Azadibenzocyclooctyne-PEG
Applications: For copper-free click chemistry reactions

Appearance: White to grey solid

Caution: Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.


Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty