Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Protease Inhibitor Cocktail V, EDTA-Free, Animal-Free

Supplier: Alfa Aesar
J64920-LQ J64920-LQ
AAJ64920-LQEA 62.43 USD InStock
AAJ64920-LQ CAAAJ64920-LQ
Protease Inhibitor Cocktail V, EDTA-Free, Animal-Free
Cell Culture Supplements and Additives
Lyophilized solid

Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized solid
Sensitivity: Hygroscopic
Appearance: White
UN No.:UN3261

Applications: For the inhibition of serine- and cysteine-proteases, but not metalloproteases. For applications that require animal-free reagents

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty