Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lipoprotein, high density, human plasma, 99%, BT-914

Supplier: Alfa Aesar
J64903-MA J64903-MA
AAJ64903-MAEA 251.38 USD InStock
AAJ64903-MA CAAAJ64903-MA
Lipoprotein, high density, human plasma, 99%, BT-914
Proteins and Peptides
Liquid

Purity: 99%
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
MDL No.: MFCD00164407

Applications: Useful in cardiovascular research and the study of lipid transport mechanisms.

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty