(±)-α-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid

Supplier: Thermo Scientific Chemicals

Synonyms: AMPA(±)-AMPAD,L-alpha-Amino-3-hydroxy-5-methylisoxazole-4-propionic acid

J64884-MA J64884-MA
AAJ64884-MAEA 297.26 USD
CAAAJ64884-MA AAJ64884-MA
(±)-α-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid
(±)-alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid

Selective quisqualate agonist


Powder

Formula: C₇H₁₀N₂O₄
MW: 186.167 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00213388
CAS Number: 74341-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR