EX 527
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Chloro-2,3,4,9-tetrahydro-1H-carbazole-1-carboxamide
, EX-527
J64753-MA
J64753-MA
AAJ64753-MAEA
153.91
USD
CAAAJ64753-MA
AAJ64753-MA
EX 527
EX 527
A potent and selective inhibitor of the SIRT1 class III histone deacetylase enzyme
Powder
Formula:
C₁₃H₁₃ClN₂O MW: 248.71 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03009471 CAS Number: 49843-98-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...