Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chymotrypsinogen A, from bovine pancreas

Supplier: Alfa Aesar
J64741-06 J64741-03 J64741-03 J64741-06
AAJ64741-03EA 126.32 USD InStock
CAAAJ64741-06 CAAAJ64741-03 AAJ64741-03 AAJ64741-06
Chymotrypsinogen A, from bovine pancreas
Proteins and Peptides
Powder

CAS No.: 9035-75-0
Formula Weight: 25.6kDa
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00130483

Applications: The zymogen of chymotrypsin

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty