5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Amino-1-(beta-D-ribofuranosyl)-1H-imidazole-4-carboxamide
J64720-MF
J64720-MF
AAJ64720-MFEA
169.4
USD
CAAAJ64720-MF
AAJ64720-MF
5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside 98%
5-Aminoimidazole-4-carboxamide-1-beta-D-ribofuranoside
Activator of AMP-activated protein kinase (AMPK)
- Powder
Powder
Formula:
C₉H₁₄N₄O₅ MW: 258.23 g/mol Melting Pt: 214…215 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00869751 CAS Number: 2627-69-2 EINECS: 220-097-5 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...